Learn More
Fisherbrand™ Economy Chemical-Resistant Goggles
SCI_ED
Catalog No. s68660a Shop All Fisherbrand Products
Click to view available options
:
Chemical Splash Goggles
This item is not returnable. View return policy
This item is not returnable. View return policy
Affordable chemical-resistant goggles.
  • Clear polycarbonate lens
  • Soft, flexible PVC body molds to facial contours for a comfortable fit
  • Adjustable headstrap ensures custom fit
  • Resist a broad range of chemicals
  • Circular vents allow air to flow freely through goggle body
Compliance
Meets ANSI/ISEA Z87.1-2015 American National Standard for Occupational and Educational Personal Eye and Face Protection Devices, Section 8.1 Droplet and Splash Hazard (D3)
Specifications
Lens Coating Anti-fog
Certifications/Compliance ANSI/ISEA Z87.1-2015
Lens Tint Clear
Product Type Chemical Splash Protection Goggles
Material Polycarbonate
Frame Material PVC
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.