Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Economy Grade Latex Coated Palm Gloves

Catalog No. p-3832400 Shop All Fisherbrand Products
Click to view available options
Size:
Small
Large
This item is not returnable. View return policy
This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
This item is not returnable. View return policy
This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.

Economical palm-coated gloves combine cost with comfort for an effective level of protection.

  • Red latex, smooth palm coated
  • Coated palm offers abrasion resistance and increased grip
  • Uncoated back allows hands to breathe
  • Ideal for assembly, construction, inspection, material handling, refuse handling, and landscaping

Specifications

Coated Area Palm
Coating Material Latex
Color Natural
Grade Economy
Material Polycotton
Product Type Dip-coated Gloves
Size Small
For Use With (Application) Assembly, Construction, Inspection, Material Handling, and Landscaping
Glove Cuff Style Continuous Knit
Series 39-C121
Length (English) 8.5 in.
Length (Metric) 21.6 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.