Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Electronic Pipetters

Catalog No. 2116584
Click to view available options
Volume (Metric):
120 μL
This item is not returnable. View return policy
This item is not returnable. View return policy

Requires

Must order carousel charging stand (sold separately).

Electronic tip ejection combined with unique ergonomic design

User-friendly design takes the effort out of pipetting and reduces the risk of work-related upper limb disorders that are common in mechanical pipetting. Choose single, 8-, and 12-channel models in a variety of volume ranges.

  • Clear LCD readout and easy-to-use keypad
  • Electronic tip-ejection buttons, located for left- or right-handed operation, eliminate tip release pressure
  • User-programmable modes, storable routine pipetting protocols, and fully adjustable speed settings for all operations
  • Electronic start button activates all piston actions, reducing strain on thumb joints and carpal tunnel Computerized drive system lets you work more efficiently than with mechanical or other electronic pipetters Replaceable tip cone filter on most models helps minimize risk of internal contamination
  • Operations include pipetting, reverse pipetting, pipetting with mixing, pipetting with cycle counter, manual pipetting, multi-dispensing, diluting, diluting with mixing, sequential dispensing, multi-aspirating, and automatic multi-dispensing.
  • Can be charged using single-pipetter charging stand, 4-pipetter carousel charging stand, or AC-charging adapter; replacement NiMH battery packs are available (FS Cat. No. 21-390-45)

Order Info

Compatible with most universal fit tips.

TRUSTED_SUSTAINABILITY

Specifications

Accuracy 2 to 0.5%
Battery Type NiMH
Compatible Tips Universal fit pipette tip
Adjustable Tip Spacing Yes
Autoclavable Not Autoclavable
Color Gray
Disposable Reusable
For Use With Universal Pipette Tip
Material Polypropylene
Number of Channels 8
Sterility Non-sterile
Type Electronic Pipette
Volume (Metric) 120 μL
Display LCD
Finger Style Ergonomic
For Use With (Equipment) Universal fit pipette tip
Increments 0.5μL
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.