Learn More
Description
Quick-drying ink offered in a variety of colors
Designed to write on damp surfaces as easily as they write on dry surfaces. Proven permanent on most surfaces, including plastic, metal, and ceramic.
- Fine tip is excellent for general marking
- Highly resistant to abrasion and water
- Will not smear when subjected to water or alcohol during normal use
Specifications
Specifications
| Product Type | Lab Marker |
| Tip Style | Fine |
| Color | Black |
| Sterility | Non-sterile |
| Latex Free | Yes |
| For Use With (Application) | Used to make permanent markings on plastic, metal, or porcelain labware |
| Quantity | 10 Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.