Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Formalin Neutralizing/Absorbing Pads
SDP

Catalog No. 2202670 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Blue
White
Packaging Quantity:
30 Pads/Cs.
55 Sheets/Roll, 6 Rolls/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Packaging Quantity
22-026-70 Blue 55 Sheets/Roll, 6 Rolls/Cs.
22-026-71 White 30 Pads/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-026-70
Only null left
Add to Cart
Add to Cart

For absorbing and neutralizing formalin in a lab environment

  • Available in individual pad and handy roll formats for ease of use and economics
  • Treated for the ultimate destruction of 10% Neutral Buffered Formalin
  • Neutralizes and reduces harmful vapors during grossing procedures
  • Blue color of sheets provides excellent contrast for tissue grossing
  • Large individual pads absorb, encapsulate, and neutralize up to 900mL of 10% Neutral Buffered Formalin
  • Small absorbent pads are supplied on a roll

Small Absorbent Pads (Sheets on Rolls)

  • 11 × 10 in. sheets
  • Super Absorbent Pads:
    • 9 × 21 in. pads
    • Pads absorb, encapsulate, and neutralize up to 900cc of 10% Neutral Buffered Formalin

    Specifications

    Color Blue
    Type Formalin Absorbent Pad
    For Use With (Application) Neutralizes and reduces harmful vapors during grossing procedures
    Packaging Quantity 55 Sheets/Roll, 6 Rolls/Cs.
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.