Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Fluorescent Antibody Microscope Slides
Shop All Fisherbrand Products
Click to view available options
Color:
White
Description:
Light white Superfrost tab slide with two 10mm diameter circles
Light white Superfrost tab slide with two 15mm diameter circles
Quantity:
72 Slides/Box, 20 Boxes/Case
Description
- Smooth marking area for effortless writing, automated printing, or printed labels
- Manufactured from pure, water-white glass
- Pre-cleaned for immediate use
- With two 15mm diameter circles
Specifications
Specifications
Length (Metric) | 75 mm |
Thickness (Metric) | 1 mm |
Edges | Ground |
Width (Metric) | 25 mm |
Material | Glass |
Type | Microscope Slide |
Frosted | Yes |
Color | White |
Corner Style | 90° |
Description | Light white Superfrost tab slide with two 15mm diameter circles |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction