Description
- Tips are heavy platinum (1.8g), riveted to stainless steel
- Straight-type forceps with grooved handles for good gripping
Price varies with platinum market.
Specifications
Specifications
| Tip Style | Straight |
| Material | Stainless Steel |
| Product Type | Forceps, General Purpose |
| Quantity (No. per Case) | Each |