Learn More
Fisherbrand™ Histosette™ II Biopsy Processing/Embedding Cassettes with Combination Lid and Base
Shop All Fisherbrand Products
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Pink
White
Yellow
Description
- Disposable, plastic cassettes hold tissue specimens during embedding, processing, and storage
- Molded from a special high-density polymer which is resistant to the chemical action of histological solvents
- Efficient flow-through slots maximize fluid exchange and ensure proper drainage
- One-piece integral lid eliminates the need for separate steel lids
- The anterior writing surface is slanted at a 45° angle, making the cassette more suitable for use with certain types of labeling instruments
Order Info
Available in 11 noncytotoxic, nonmetallic colors.
Specifications
Specifications
Pore Style | Slots |
Writing Face Angle | 45° |
Closure Type | Combination Lid |
Color | Gray |
Material | Plastic |
Product Type | Biopsy Cassette |