Learn More
Fisherbrand™ Histosette™ II Biopsy Processing/Embedding Cassettes with Separate Lid and Base
Shop All Fisherbrand Products
Click to view available options
Color:
Aqua
Gray
Green
Orange
Peach
Pink
Tan
White
Description
- Disposable, plastic cassettes hold biopsy specimens during embedding, processing, and storage
- Molded from a special high-density polymer which is resistant to the chemical action of histological solvents
- Efficient flow-through slots maximize fluid exchange and ensure proper drainage
- The anterior writing surface is slanted at a 45° angle, making the cassette more suitable for use with certain types of labeling instruments
Order Info
Available in 11 noncytotoxic, nonmetallic colors.
Specifications
Specifications
| Pore Style | Slots |
| Writing Face Angle | 45° |
| Closure Type | Separate Lid |
| Color | Pink |
| Material | Plastic |
| Product Type | Biopsy Cassette |