Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ HMIG and NFR Write-On Labels


Shop All Fisherbrand Products



Click to view available options
Display Text:
HMIG
NFR
Description
- HMIG paper labels are large enough to include the chemical name, common name and manufacturer name
- NFR labels let you write in the chemical name, owner, date and hazard rating
Order Info
Labels measure 3.87L x 1.5 in. W (9.8 x 3.8cm).
Specifications
Specifications
| Format | Label |
| Product Type | HMIG Write-On™ Label |
| Display Text | HMIG |
| For Use With (Application) | Ideal for adding vital hazard information to surfaces of small bottles, cans and other containers |
| Quantity | 1000/Pk. |
| Length (English) | 3.87 in. |
| Length (Metric) | 9.8 cm |
| Width (English) | 1.5 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction