Learn More
Fisherbrand™ Infecon™ 3000 Infectious Substance Shipper
Click to view available options
Type:
Bubble Pouch Absorbent Strips
Bubble Wrap and Absorbent Strips
Infectious Substance Shipper
Pressure Vessels
Refurbishment
Description
- Each shipper consists of a transparent pressure vessel able to withstand up to 95kPa, cushioning materials, outer box, labels, declarations and instructions
- Transparent pressure vessel allows contents to be seen
- Use the Infecon 3000 with the Speci-Freez™ Insulated Shipper (19-075-384C) for refrigerated or frozen specimens
- Reusable when using the complete refurbishment kit (19-037-437)
Order Info
Dimensions: 6 x 6 x 8 in. (15.24 x 15.24 x 20.32cm) with 1L vessel
Specifications
Specifications
| Applications | For infectious substances |
| Type | Bubble Pouch Absorbent Strips |