Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Lab Automation Pipet Tips
GREENER_CHOICE

Catalog No. p-159814 Shop All Fisherbrand Products
Click to view available options
Volume (Metric):
1 to 20 μL
100 μL
200 μL
250 μL
5 to 250 μL
50 μL
Sterility:
Non-sterile
Sterile
Color:
Blue
Natural

Provide maximum performance, accuracy and reproducibility in laboratory automation procedures

Carbon-black, liquid-level-sensing tips are required by many automated systems. They are constructed of a highly conductive material that is necessary to detect presence or absence of sufficient liquid, in order to avoid short sampling and excessive submersion of tip during aspiration.

Disposable tips for robotic workstations — provide maximum performance, accuracy, and reproducibility in laboratory automation procedures. Available in clear, blue tint, or carbon-black-conductive polypropylene.
Features

  • Ideal for demanding HTS procedures
  • Provide maximum performance, accuracy, and reproducibility in lab automation procedures
  • Engineered to ensure proper fit and function with several standard and customized automated liquid-handling systems
  • Choose tips in an assortment of volume sizes and styles

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1 to 20 μL
Sterility Non-sterile
Color Natural
Autoclavable Autoclavable
For Use With (Application) Ideal for demanding HTS procedures, provide maximum performance, accuracy and reproducibility in lab automation procedures
Format Rack
Tip Style Automation
Disposable Single-use
For Use With (Equipment) Beckman Coulter Biomek 1000/2000
Material Polypropylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.