Learn More
Fisherbrand™ Multipurpose Specimen Storage Containers
SDP
Catalog No. 02544216B Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (English):
4 oz.
Catalog No. Capacity (English)
02-544-216B 4 oz.
1 options
This item is not returnable. View return policy
Catalog No. 02-544-216B
This item is not returnable. View return policy
Ideal for storing biologically hazardous samples for autoclaving before disposal
  • Molded of high-density polyethylene
  • Inert to most chemicals including formaldehyde, weak acids and all bases
  • Resistant to freezing and boiling
  • Stackable and virtually indestructible
  • Easy to write on with either pen or pencil
  • Not graduated
Specifications
Capacity (English) 4 oz.