Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Crinkle Latex Coated Seamless Knit Cotton/Polyester Gloves

Catalog No. 19816907 Shop All Fisherbrand Products
Click to view available options
Size:
Medium
Large
X-Large
This item is not returnable. View return policy
This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
This item is not returnable. View return policy
This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.

Ideal for assembly, construction, inspection, and general material handling.

  • Seamless cotton-polyester liner with crinkle-textured latex palm coating
  • Enhances wet and dry grip, while also repelling moisture from skin
  • Latex coating on the palm, fingertips, and thumb crotch ensures longer wear
  • Ergonomic shape reduces hand fatigue
  • Machine washable

Specifications

ANSI Abrasion Rating ANSI 3
Coated Area Palm, Fingers, Thumb Crotch
Construction Seamless Knit
Coating Material Crinkled Latex
Color Blue, Gray
Exterior Finish Fully Textured
Grade Economy
Hand Specific
Material Polycotton
Product Type Seamless Knit Gloves
Size Large
EN388 Standard 2142
For Use With (Application) Assembly, Construction, Inspection, Material Handling, and Landscaping
Glove Cuff Style Continuous Knit
Series 39-1310
Length (English) 10.3 in.
Length (Metric) 26.1 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.