Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Nonsterile Cotton Balls
SDP

Catalog No. p-4653903 Shop All Fisherbrand Products
Click to view available options
Weight (Metric):
0.004 kg
0.45 kg
1.08 kg
Quantity:
2000 Cs.
4000 Cs.
Size:
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Use these cotton balls for a wide range of applications. Fisherbrand™ Nonsterile cottonballs are available in a variety of sizes.

  • Uniform in size and shape
  • Made of pure, natural, bleached cotton for increased absorbency
  • Available in sizes medium, large, extra large and bulk roll

Order Info

For the sterile version of 22-456-880, please order 22-456-885.

Specifications

Product Type Cotton Balls
Sterility Non-sterile
Size Medium
Quantity 4000 Cs.
Weight (English) 0.006 lb.
Weight (Metric) 0.45 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.