Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Permaloc™ Formalin Containers
SDP

Catalog No. p-4881975 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
120 mL, pre-filled 60 mL
20 mL, pre-filled 10 mL
40 mL, pre-filled 20 mL
60 mL, pre-filled 30 mL
90 mL, pre-filled 45 mL
No. per Case:
128/Cs.
192
324/Cs.
48/Cs.
480/Cs.
768/Cs.

Catalog No. 22-126-347

Double-click locking ensures specimen integrity while guarding against lost specimens through accidental spills

  • Patented double-clicking lock design
  • 1st audible“click”ensures closure
  • 2nd audible“click”achieves full 95kPa certification
  • Available in two convenient sizes –20mL (10mL filled) neutral-buffered formalin;60mL (30mL filled) with wide mouth (53mm) opening neutral-buffered formalin
  • Leak tested and independently 95kPa certified
  • Avoid employee/patient exposure to hazardous chemicals
  • Improve efficiency of specimen transport to the laboratory
  • Economical bulk packaging with inner convenient transport packs help grow your Outreach Programs

Specifications

Product Type Formalin Container
Capacity (Metric) 20 mL, pre-filled 10 mL
Closure Type Double clicking lock design
No. per Case 768/Cs.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.