Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Permaloc™ Formalin Containers
Shop All Fisherbrand Products

Click to view available options
Capacity (Metric):
120 mL, pre-filled 60 mL
20 mL, pre-filled 10 mL
40 mL, pre-filled 20 mL
60 mL, pre-filled 30 mL
90 mL, pre-filled 45 mL
No. per Case:
128/Cs.
192
324/Cs.
48/Cs.
480/Cs.
768/Cs.
Description
- Patented double-clicking lock design
- 1st audible“click”ensures closure
- 2nd audible“click”achieves full 95kPa certification
- Available in two convenient sizes –20mL (10mL filled) neutral-buffered formalin;60mL (30mL filled) with wide mouth (53mm) opening neutral-buffered formalin
- Leak tested and independently 95kPa certified
- Avoid employee/patient exposure to hazardous chemicals
- Improve efficiency of specimen transport to the laboratory
- Economical bulk packaging with inner convenient transport packs help grow your Outreach Programs
Specifications
Specifications
Product Type | Formalin Container |
Capacity (Metric) | 20 mL, pre-filled 10 mL |
Closure Type | Double clicking lock design |
No. per Case | 128/Cs. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction