Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Unlined Pigskin Leather Drivers Gloves

Catalog No. 19043030 Shop All Fisherbrand Products
Click to view available options
Size:
Medium
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Gloves return to their original pliability even after being soaked in water.

  • Suitable for construction, machine operation, utility work and many general applications
  • Gunn-cut style with straight thumb
  • Elastic wrist provides a sure fit
  • Unlined

Specifications

Color Natural
Material Pigskin Leather
Product Type Driver Gloves
Size Medium
Style Gunn
For Use With (Application) Construction, Machine Operation, Utility Work and General Applications
Glove Cuff Style Open
Hem Color Green
Thumb Style Straight
Length (English) 9.6 in.
Length (Metric) 24.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.