Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Polypropylene Disposable Beakers
Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
100 mL
Description
- Highly resistant to strong acids, organic solvents and other common laboratory reagents
- Special “dripless” lip Rib construction permits stacking without binding Base allows beaker to sit flat during use
- Graduated in mL and oz.
Specifications
Specifications
Capacity (English) | 3.38 oz. |
Capacity (Metric) | 100 mL |
Material | Polypropylene |
For Use With (Application) | Highly Resistant to Strong Acids, Organic Solvents and Other Common Laboratory Reagents |
Includes | Spout |
Increments | 50 mL |
Type | Beaker |
Autoclavable | Autoclavable |
Disposable | Single-use |
Graduated | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction