Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Porcelain Spot Plates
SCI_ED

Catalog No. s02597 Shop All Fisherbrand Products
Click to view available options
No. of Wells:
12
This item is not returnable. View return policy
This item is not returnable. View return policy

For observing color reactions

  • Material: porcelain
  • Plate color: white
  • Available with 12 wells

Specifications

Type Spot Plate
Dimensions (L x W) 4.7 x 3.6 in. (118 x 91 mm)
No. of Wells 12
For Use With (Application) For observing color reactions
Color White
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.