Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ PTFE Disposable Stir Bar
Shop All Fisherbrand Products

Click to view available options
Length (Metric):
25.4 mm
41.3 mm
50.8 mm
Length (English):
1 in.
1.63 in.
2 in.
Description
- Ideal for single use applications mixing radioactive reagents or when cross contamination might be a concern
- Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE low friction coating; durable and inert
- Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
- Autoclavable
Specifications
Specifications
| Coating | PTFE, FDA-grade, USP Class VI |
| Shape | Polygon |
| Type | Magnetic Stir Bar |
| Material | Alnico Magnet, PTFE |
| Autoclavable | Autoclavable |
| Diameter (Metric) | 8 mm |
| Length (Metric) | 41.3 mm |
| Quantity | 100 |
| Certifications/Compliance | ISO 9001:2008 |
| Color | White |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction