Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Seamless Knit Cotton/Polyester Gloves with Double-Sided PVC Dot Grip

Catalog No. p-144503 Shop All Fisherbrand Products
Click to view available options
Size:
Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Used in warehousing and package handling, general assembly, light duty general work situations, construction, and bottle handling.

  • Seamless knit shell is comfortable and flexible
  • Knit Wrist helps prevent dirt and debris from entering the glove
  • Double-sided PVC dots enhance grip and provide additional resistance to cuts and abrasions
  • Small size fits ladies well

Specifications

Construction Seamless Knit
Coating Material PVC Dots (2 Sides)
Color Natural, Blue
Cut Resistance Rating ANSI A3
Hand Ambidextrous
Material Polycotton
Product Type Seamless Knit Gloves
Size Large
For Use With (Application) Light Duty Tasks, Bottle Handling, Construction, General Assembly, and Warehousing
Glove Cuff Color Natural
Glove Cuff Style Knit
Series 37-C110DD
Length (English) 10 in.
Length (Metric) 25.4 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.