Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ String Knit Gloves with PVC Dot Grip

Catalog No. p-144487 Shop All Fisherbrand Products
Change view
Click to view available options
Coating Material:
PVC Dots (1 Side)
PVC Dots (2 Sides)
Size:
Small
Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Coating Material Size
11-387-57 PVC Dots (1 Side) Small
19-816-783 PVC Dots (1 Side) Large
11-387-52 PVC Dots (2 Sides) Small
11-387-53 PVC Dots (2 Sides) Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 11-387-57
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

PVC dots enhance grip capabilities and abrasion resistance.

  • Cotton string knit gloves with PVC dots on either one side or both sides
  • Ideal for protecting hands from dirt and abrasion
  • Seamless construction is comfortable and reversible
  • Available in universal women's and men's sizes
  • Color-coded hem for quick size identification
  • Standard weight
  • Launderable

Specifications

Construction String Knit
Accelerator Free No
Applications Industrial
Coating Material PVC Dots (1 Side)
Color Natural, Black
Cut Resistance Rating ANSI A1
Disposable Reusable
Exterior Finish Grip Pattern
Hand Specific
Material Cotton
Powdered Powder Free
Product Type String Knit Gloves
Size Small
Sterility Non-sterile
Glove Cuff Style Continuous Knit
Grip Design Dots
Hem Color Yellow
Length (English) 8.9 in.
Length (Metric) 22.6 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.