Learn More
Fisherbrand™ Racer Series Qualifier Eyewear
Offers exceptional protection and style at an affordable price
 View more versions of this product
Catalog No. 19-042-1001
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
This item is not returnable. View return policy
Description
Description
Offers exceptional protection and style at an affordable price
  • Flared single-lens design provides sleek protection
  • Soft nose bridge and bayonet-style side arms grip lightly but firmly for a comfortable, confident fit
  • Black nylon frames
Compliance
Meets ANSI* Z87.1 and Z80.3 standards.
Specifications
Specifications
Scratch-resistant
Dark Gray
Polycarbonate
ANSI Z87.1, ANSI Z80.3
Black
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.