Learn More
Fisherbrand™ Racer Series Qualifier Sport Eyewear
Frameless single lens design with soft nose bridge and temples to grip the face lightly but firmly
This item is not returnable.
View return policy
Description
- Frameless single lens design
- Soft nose bridge and temples to grip the face lightly but firmly
- Blue lens commonly used for indoor applications with an excessive amount of yellow or sodium vapor light
- Gray lens minimizes outdoor sunlight and glare Clear lens is excellent for most indoor applications
Compliance
Meets ANSI* Z87.1 and Z80.3 standards.
Specifications
ANSI Z87.1, ANSI Z80.3 | |
Clear | |
Black, Blue |