Learn More
Fisherbrand™ Racer Series Qualifier Sport Eyewear
Frameless single lens design with soft nose bridge and temples to grip the face lightly but firmly
Catalog Number | Frame Color | Lens Tint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Frame Color | Lens Tint | Price | Quantity & Availability | |||||
19-181-540
|
Blue | Blue | N/A | N/A | N/A | ||||
19-181-542
|
Silver, Black | Gray | N/A | N/A | N/A | ||||
19-181-541
|
Black, Blue | Clear | N/A | N/A | N/A | ||||
Description
- Frameless single lens design
- Soft nose bridge and temples to grip the face lightly but firmly
- Blue lens commonly used for indoor applications with an excessive amount of yellow or sodium vapor light
- Gray lens minimizes outdoor sunlight and glare Clear lens is excellent for most indoor applications
Compliance
Meets ANSI* Z87.1 and Z80.3 standards.
Specifications
ANSI Z87.1, ANSI Z80.3 |