Description
- High-visibility unilens provides maximum field of vision
- A soft nose bridge and rubber-tipped side arms offer a comfortable grip
Compliance
Meets ANSI* Z87.1 and Z80.3 standards.
Specifications
Specifications
| Lens Coating | Scratch-resistant |
| Certifications/Compliance | ANSI Z87.1, ANSI Z80.3 |
| Lens Tint | Clear |
| Frame Color | Silver |
| Lens Material | Polycarbonate |