Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Reusable Polyethylene Dippers

500mL beakers with long handles for removing samples from large tanks.

 View more versions of this product

Catalog No. 14-242-1


Add to cart

Description

Description

500mL beakers with long handles for removing samples from large tanks.

Order Info

Additional dipper sizes available. To order, contact your Fisher Scientific Customer Service Representative.

Specifications

Specifications

16.9 oz., 500mL
For removing samples from large tanks
109.22cm
5.75 in.
14.6cm
5 in.
16.9 oz.
Dipper
5.75 x 5 x 43 in. (14.6 x 12.7 x 109.22cm)
43 in.
3 ft.
0.9m
HDPE
12.7cm
500mL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit