Learn More
Fisherbrand™ Ringed Cytology Slides
Shop All Epredia Products
Click to view available options
Type:
Double Ringed Cytology Slide
Double Ringed with Adhesive Cytology Slide
Single Ringed Cytology Slide
Single Ringed with Adhesive Cytology Slide
Description
- Fully compatible with Fisherbrand and Epredia™ Shandon™ Cytology Funnels
- Available with single or double rings, with or without adhesive coating
Specifications
Specifications
Type | Double Ringed with Adhesive Cytology Slide |
Description | Round |
For Use With (Application) | For cytology funnels, designed to collect a thin layer of cells in the ringed area |
Dimensions (L x W) | 13 mm |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov