Learn More
Fisherbrand™ Safety Blue Baume (Heavy) Thermohydrometers
SDP
Catalog No. p-4704738 Shop All Fisherbrand Products
Click to view available options
Baume Range:
0 to 12
19 to 31
29 to 41
35 to 70
39 to 51
49 to 61
59 to 71
9 to 21
Catalog No. 11-555-242
Only null left
Add to Cart
Add to Cart
Lead and mercury free thermohydrometers.
Hydrometer with enclosed thermometer to indicate temperature of the liquid under test.
TRUSTED_SUSTAINABILITY
Specifications
Increments 0.1
Certifications/Compliance NIST
Length (Metric) 385 mm
Material Soda Lime Glass
Ballast Type Lead and Mercury Free
Baume Range 39 to 51
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.