Learn More
Description
- An efficient device for crossmatching blood type for transfusion
- Large labels feature easy-to-read, white information area, pre-printed serial numbers and “carbonless copy“ underneath
- Labels are removed from the band and adhered to draw tubes, units of blood, other specimen tubes or paperwork
- Match the serial numbered label on the unit of blood with the patient's blood band before transfusion to prevent fatal errors
Specifications
Specifications
| Includes | Large labels |
| Color | Yellow |
| Product Type | Securline™ Blood Band |
| For Use With (Application) | Trauma centers, efficient device for crossmatching Blood type for transfusion |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.