Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Securline™ Blood Bands

Catalog No. 1490519 Shop All Fisherbrand Products
Click to view available options
Color:
Red
Yellow

Ideal for use in trauma centers

  • An efficient device for crossmatching blood type for transfusion
  • Large labels feature easy-to-read, white information area, pre-printed serial numbers and “carbonless copy“ underneath
  • Labels are removed from the band and adhered to draw tubes, units of blood, other specimen tubes or paperwork
  • Match the serial numbered label on the unit of blood with the patient's blood band before transfusion to prevent fatal errors

Specifications

Includes Large labels
Color Red
Product Type Securline™ Blood Band
For Use With (Application) Trauma centers, efficient device for crossmatching Blood type for transfusion
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.