Learn More
Description
Puncture-resistant with wide rectangular opening for horizontal loading of needles
- General-purpose sharps container with clear locking lid and marked with biohazard label
- Made from puncture-resistant polypropylene that can be autoclaved or incinerated
- Easily mount on walls and carts, nest to conserve storage space
Compliance
Used to comply with OSHA standards for sharps and biohazard waste collection.
Specifications
Specifications
| Capacity (Metric) | 26.5 L |
| Certifications/Compliance | Used to comply with OSHA standards for sharps and biohazard waste collection. |
| Closure Type | Lockable |
| Closure Color | Clear |
| Capacity (English) | 7 gal. |
| Color | Red |
| Dimensions (L x W x H) | 12 x 15.5 x 14 in. |
| Disposable | Single-use |
| Height (Metric) | 36 cm |
| Length (Metric) | 31 cm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.