Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Shipping Labels

Catalog No. 19038302E Shop All Fisherbrand Products
Click to view available options
Product Type:
Cargo Aircraft Only Label
Dry Ice Mini Super Label
Inner Packaging Complies Label
Substance Label
Size (English):
2 x 2 in.
4 x 2.75 in.
4.27 x 4.5 in.
6 x 6 in.
Size (Metric):
10.2 x 7 cm
10.84 x 11.43 cm
15.24 x 15.24 cm
5 x 5 cm
This item is not returnable. View return policy
This item is not returnable. View return policy

For shipping infectious substances and diagnostic specimens

Specifications

Format Label
Product Type Inner Packaging Complies Label
For Use With (Application) Shipping
Quantity 12/Pk.
Size (English) 4 x 2.75 in.
Size (Metric) 10.2 x 7 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.