Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ SHUR/Clean™ Active Sloped-Front Fume Hoods for Coverslipping
SDP

Bench-top fume hoods to help manage routine manual histological processes

$7790.00

Specifications

Product Type Ductless Fume Hood
Depth (English) 20 in.
Depth (English) Interior 19 in.
Depth (Metric) 51 cm
Depth (Metric) Interior 48 cm
View More Specs

Products 2
Catalog Number Voltage Price Quantity & Availability  
Catalog Number Voltage Price Quantity & Availability  
22-047-203
SDP
View Documents
100/115 V
Each for $7,790.00
Only null left
Add to Cart
 
22-047-204
SDP
View Documents
220/240 V
Each for $7,790.00
Only null left
Add to Cart
 
Description

Description

The Sloped-Front Fume Hood features two 174 cfm high-speed fans that pull contaminated air through two 8 x 8 in. prefilters to remove dust and particulate matter and two 8 x 8 in. charcoal filters to provide the protection needed for hazardous fume absorption. Double-hinged front sash provides convenient access for multiple applications.

  • Microprocessor controlled
  • Double-hinged sash
  • Two prefilters for particulate removal and two carbon filters for fume containment
  • Two constant-speed 174 cfm blowers
  • Face velocity: 90 fpm with sash completely down
  • 22 in. fluorescent 13W T-5 bulb
  • External dimensions: 36W x 20D x 28 in.H (91 x 51 x 71cm)
  • Internal dimensions: 34W x 19D x 19 in.H (86 x 48 x 48cm)
  • Weight: 67 lb. (31kg)
  • Model SC-36S-X (for Coverslipping): contains two carbon filters (SC-36S-XF) for xylene fume containment

Specifications

Specifications

Ductless Fume Hood
19 in.
48 cm
Coverslipping
19 in.
48 cm
31 kg
34 in.
86 cm
20 in.
51 cm
19 x 34 x 19 in. (48 x 86 x 48 cm)
28 in.
71 cm
67 lb.
36 in.
91 cm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.