Learn More
Fisherbrand™ SHUR/Clean™ Passive Open-Front Fume Hoods

Without filters for installations to be externally exhausted
Specifications
Depth (English) | 26 in. |
---|---|
Depth (Metric) | 66 cm |
Height (English) | 28 in. |
Height (Metric) | 71 cm |
Catalog Number | Exhaust | Width (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Exhaust | Width (Metric) | Price | Quantity & Availability | |||||
22-047-207
![]() |
91 cm | N/A | N/A | N/A | |||||
22-047-208
![]() |
91 cm | N/A | N/A | N/A | |||||
22-047-209
![]() |
122 cm | N/A | N/A | N/A | |||||
22-047-210
![]() |
122 cm | N/A | N/A | N/A | |||||
Description
Exhaust ports are on the top or rear mounted. Can easily accomodate Microwave Tissue Processor and its accessories. (Contact your local architect and/or mechanical contractor to determine what accessories will be required to comply with local building codes.)
Requires ducting to HVAC system by user
Top (-TV) or rear (-RV) 6 in. O.D. exhaust ports
- External dimensions: 36W x 26D x 28 in.H (91 x 66 x 71cm)
- Internal dimensions: 34 x 25 x 27 in. (86 x 64 x 69cm)
- Weight: 26 lb. (12kg)
- Accommodates the following number of reagent bowls when used with Microwave Tissue Processor and optional Reagent Bowl Shelf (22047211): 4 large, 4 med. or 4-6 small bowls
- External dimensions: 48W x 26D x 28 in.H (122 x 66 x 71cm)
- Internal dimensions: 46 x 25 x 27 in. (117 x 64 x 69cm)
- Weight: 32 lb. (15kg)
- Accommodates the following number of reagent bowls when used with Microwave Tissue Processor and optional Reagent Bowl Shelf (22047212): 8 large, 8 med. or 10 small bowls
Specifications
26 in. | |
66 cm | |
28 in. | |
71 cm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.