Learn More
Fisherbrand™ Stainless-Steel Pressure Vessels Accessories
Shop All Fisherbrand Products
Click to view available options
Type:
Elbow Fitting
Hose Connector
Housing Plug
Pressure Gauge
Description
- A range of gauges, fittings and adapters for pressure vessels
Specifications
Specifications
Connectors | 1/4 in. NPTM |
For Use With (Equipment) | Pressure Vessel |
Material | Stainless Steel |
Max. Pressure | 160 psi |
Type | Pressure Gauge |