Learn More
Fisherbrand™ Superfrost™ Plus Microscope Slides PROMO
Catalog No. 1255015S24 Shop All Fisherbrand Products
Click to view available options
Corner Style:
Clipped
Regular
This item is not returnable. View return policy
Catalog No. 12-550-15S24
This item is not returnable. View return policy
Superfrost™ Plus Microscope Slides are an ideal choice for frozen sections, cytocentrifuge preparations, and standard Papanicolaou smears.
  • Maintain the same features as regular Superfrost™ slides with a special treatment process that electrostatically adheres tissue sections and cells to the glass without the need for adhesives or protein coatings
  • Treatment virtually eliminates tissue loss during staining
  • Saves hours of slide preparation
  • Chemical-resistant, white marking surface
  • 3/4 in. Superfrost™ tab
Specifications
Length (Metric) 75 mm
Width (Metric) 25 mm
Frosted 1 End
Type Microscope Slide
Material Glass
Color White
Corner Style Regular
Quantity 72 Slides/Pk., 2 Pk./Cs.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.