Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Superfrost™ Slides

Catalog No. p-3574518 Shop All Fisherbrand Products
Click to view available options
Color:
Clear
Green
Length (Metric):
75 mm
Width (Metric):
25 mm
50 mm
Quantity:
144 Pk., 10 Pk./Cs.
144/Pk., 5 Pk./Cs.
This item is not returnable. View return policy
This item is not returnable. View return policy

Durable and opaque

  • Autoclavable
  • Available in an array of tab colors
  • Offered in standard and large sizes, for positioning larger specimens
  • Durable, opaque tab is inert to common laboratory chemicals and reagents
  • Packaged in paperboard box

Specifications

Length (Metric) 75 mm
Frosted SuperFrost
Thickness (Metric) 1 mm
Width (Metric) 25 mm
Color Clear
Length (English) 3 in.
Quantity 144 Pk., 10 Pk./Cs.
Width (English) 1 in.
Dimensions (L x W) 3 x 1 in. (75 x 25 mm)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.