Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ SURE-TEK™ 2 Processing Embedding Cassettes Dispenser Packs
SDP

Catalog No. 22048144 Shop All Fisherbrand Products
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Peach
Pink
White
This item is not returnable. View return policy
This item is not returnable. View return policy

Comes with attached lids

  • Supplied in Dispenser Packs
  • Available in White, Blue, Green, Grey, Pink, Peach, Yellow, Tan, Aqua and Lilac colors

Specifications

For Use With (Equipment) Sure-tek™ 2 Cassette
Includes Attached lid
Type Processing and Embedding Cassette
Color Gray
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.