Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Healthcare™ SURE-TEK™ 2 Processing Embedding Cassettes Dispenser Packs
Shop All Fisherbrand Products

Click to view available options
Color:
Aqua
Blue
Green
Lilac
Peach
Pink
White
Description
- Supplied in Dispenser Packs
- Available in White, Blue, Green, Grey, Pink, Peach, Yellow, Tan, Aqua and Lilac colors
Specifications
Specifications
| For Use With (Equipment) | Sure-tek™ 2 Cassette |
| Includes | Attached lid |
| Type | Processing and Embedding Cassette |
| Color | Aqua |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction