Learn More
Description
- Pure cellulose paper
- Impermeable polyethylene backing
- Smooth, firm surface
- Does not inhibit stability of bottles or tubes, and allows recovery of dry spills
- Disposal is safe and easy
- Polyshield ashes readily upon incineration
Liquid samples can be recovered by washing the absorbed spill from the inert surface with the appropriate buffer.
Specifications
Specifications
| Product Type | Bench Protector |
| Material | Cellulose/Polyethylene |
| Length (English) | 164 ft. |
| Length (Metric) | 50 m |
| Width (English) | 36 in. |
| Width (Metric) | 91 m |
| For Use With (Application) | Polyshield fluid barrier is a super absorbent for splatters and spills, and protects work areas from contamination and damage by toxic, infectious, corrosive, radioactive or staining materials |
| Quantity | Each |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.