Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM), NIST tested, Fisherbrand™

Catalog No. 06664265 Shop All Fisherbrand Products
Click to view available options
Conductivity:
1 μS
10 μS
10,000 μS
100 μS
100,000 μS
1000 μS
1413 μS
150,000 μS
200,000 μS
5 μS
Resistivity:
0.000005 μ℧
0.000006 μ℧
0.00001 mOhm
0.0001 mOhm
0.00071 mOhm
0.001 mOhm
0.01 mOhm
0.1 mOhm
0.2 mOhm
0.5 mOhm
1.0 mOhm
150000 μ℧
200000 μ℧
Concentration or Composition Notes:
NIST Batch Tested
NIST Individual Bottle Tested

Use NIST-traceable reference materials for the highest accuracy available. Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM) offer easy calibration with included instructions and temperature chart.

  • Each bottle includes a NIST-traceable calibration report supplied by the manufacturer and information from batch testing
  • Also includes step-by step calibration instructions and temperature compensation chart
  • Accuracy at 25°C is (±0.25 microsiemens for 1, 5, 10 microsiemen solution or ±0.25% for other solutions) or the uncertainty shown on the certificate, whichever is greater
  • ISO 17034 (1750.02) (formerly ISO Guide 34) Certified Reference Material

Order Info

473 mL (16 oz.) Bottle

Specifications

Chemical Name or Material Conductivity Standard
Description NIST Batch Tested, Resistivity: 150000 μ℧
Packaging Bottle
Concentration or Composition Notes NIST Batch Tested
Traceability to NIST NIST Batch Tested
Conductivity 150,000 μS
Resistivity 150000 μ℧
Technique Calibration of Conductivity Instrument
Certifications/Compliance ISO 17034
Color Clear
For Use With (Equipment) Conductivity Meter
Solubility Soluble in water
Solution Type Calibration Solution
Type Conductivity Standard
Quantity 500 mL
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.