Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM), NIST tested, Fisherbrand™
Shop All Fisherbrand Products
Click to view available options
Conductivity:
1 μS
10 μS
10,000 μS
100 μS
100,000 μS
1000 μS
1413 μS
150,000 μS
200,000 μS
5 μS
Resistivity:
0.000005 μ℧
0.000006 μ℧
0.00001 mOhm
0.0001 mOhm
0.00071 mOhm
0.001 mOhm
0.01 mOhm
0.1 mOhm
0.2 mOhm
0.5 mOhm
1.0 mOhm
150000 μ℧
200000 μ℧
Concentration or Composition Notes:
NIST Batch Tested
NIST Individual Bottle Tested
Description
Use NIST-traceable reference materials for the highest accuracy available. Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM) offer easy calibration with included instructions and temperature chart.
- Each bottle includes a NIST-traceable calibration report supplied by the manufacturer and information from batch testing
- Also includes step-by step calibration instructions and temperature compensation chart
- Accuracy at 25°C is (±0.25 microsiemens for 1, 5, 10 microsiemen solution or ±0.25% for other solutions) or the uncertainty shown on the certificate, whichever is greater
- ISO 17034 (1750.02) (formerly ISO Guide 34) Certified Reference Material
Order Info
473 mL (16 oz.) Bottle
Specifications
Specifications
Chemical Name or Material | Conductivity Standard |
Description | NIST Individual Bottle Tested, Resistivity: 1.0 mΩ |
Packaging | Bottle |
Concentration or Composition Notes | NIST Individual Bottle Tested |
Traceability to NIST | NIST Individual Bottle Tested |
Conductivity | 1 μS |
Resistivity | 1.0 mOhm |
Technique | Calibration of Conductivity Instrument |
Certifications/Compliance | ISO 17034 |
Color | Clear |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction