Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
Traceable™ One-Shot Conductivity Standards, Fisherbrand™
Shop All Fisherbrand Products
                    Click to view available options
                
                
                    
                            
                                Concentration:
                            
                        
                        
                            
                                150,000 μS/μ℧
                            
                        
                            
                                200,000 μS/μ℧
                            
                        
                    Description
Eliminates contamination with calibration within disposable container. Traceable™ One-Shot Conductivity Standards are 100% compatible with all conductivity meters.
- Nominal Value: 150000 Micro (mhos/siemens)
 - Certified Reference Material
 - Accuracy: certificate stated uncertainty, or ±0.25 microsiemen or ±0.25%
 - Includes: 6-pack 100 mL container, step-by-step calibration instructions, individual temperature compensation charts, traceability information, Traceable Certificate
 
Specifications
Specifications
| Chemical Name or Material | One-Shot Conductivity Standards | 
| Packaging | Bottle | 
| CAS | 7447-40-7 | 
| Molecular Formula | ClK | 
| Concentration | 150,000 μS/μ℧ | 
| Technique | Calibration of Conductivity Instrument | 
| MDL Number | MFCD00011360 | 
| Color | Clear | 
| For Use With (Equipment) | Conductivity Meters and Probes | 
| Solubility | Soluble in water | 
| Show More | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction