Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Three-hole Twistit™ Rubber Stoppers
SDP

Catalog No. 14131H Shop All Fisherbrand Products
Click to view available options
Color:
Black
White
Stopper Number:
5
6
7
8
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Stoppers made from soft, resilient, natural rubber with safe convenient countersunk holes, stoppers feature excellent aging characteristics with high resistance to deformation

  • Simple twist-off seals provide convenient one, two, or three-hole use
  • Solid stoppers are vacuum-tested at 30 in. of mercury
  • High resistance to deformation—even after repeated autoclaving
  • Each stopper has its size number in raised figures on top

Specifications

Product Type Stopper (3 holes)
Stopper Number 10
Size 10
Total Positions 3
Material Natural Rubber
Color Black
For Use With (Application) Multi-purpose applications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.