Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Ultrasonic Units Cooling Coils

The Fisherbrand™ Ultrasonic Unit Cooling Coil is designed specifically for use with the Fisherbrand™ 112xx Series of dual frequency ultrasonic cleaners. Maintaining a stabile liquid temperature is critical in ultrasonic cleaning.
$563.20 - $675.40
Specifications
Type | Cooling Coil |
---|---|
Diameter (Metric) Inner | 6 mm |
Diameter (Metric) | 8 mm |
Material | Stainless Steel |
Description
- Keep the cooling bath temperature stable
- Ensure proper ultrasonic cleaning with the exact temperature
- Connects to the closed cycle cooling system
Specifications
Cooling Coil | |
6 mm | |
8 mm | |
Stainless Steel |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title