Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ 10-SG Urine Reagent Strips

Catalog No. 23111262 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 Strips/Canister, 24 Canisters/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-111-262 100 Strips/Canister, 24 Canisters/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-111-262
  • PRODUCT PROMOTION
    $202.00
    $31.11 / Pack of 100

    Save $170.89

    85% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • PRODUCT PROMOTION
    $3,909.00
    $746.64 / Case of 24 PK

    Save $3,162.36

    81% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For the semi-quantitative detection of the following analytes in urine GLUC, BILI, KET, SG, Blood, pH, Protein, URO, Nitrite and Leukocytes. Professional, in vitro diagnostic use. A substantial equivalent to the leading national brand. Use with the Fisherbrand Urine Analyzer or read visually.

A CLIA Waived, easy-to-use system that provides fast, highly-accurate results for the general evaluation of health, aiding in the diagnosis and monitoring of metabolic or systemic diseases that affect kidney function, endocrine disorders and diseases or disorders of the urinary tract.

  • Strips combinations with up to 10 parameters
  • Analytical sensitivity comparable to the market leader
  • High quality color chart ensures accurate visual reading
  • 2 year shelf life for unopened canisters
  • 3 month shelf life for strips in opened canisters

† For visual read or on AimStrip Analyzers.

Specifications

Content And Storage 15°C to 30°C (59°F to 86°F, room temperature)
CE Marker No
Clia Complexity Waived
Detectable Analytes Glucose, Bilirubin, Ketone, Specific Gravity, Blood, pH, Protein, Urobilinogen, Nitrite, Leukocyte
Sample Type Urine
For Use With (Application) Urinalysis
Quantity 100 Strips/Canister, 24 Canisters/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.