Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Vial Screw Caps
Shop All Fisherbrand Products
Click to view available options
Color:
Blue
Red
Description
- Made of polypropylene
- With lip seal design for leakproof seal
- Autoclavable
- For externally threaded tubes
- Tubes sold separately
Specifications
Specifications
Color | Blue |
Material | Polypropylene |
Septum | No |
For Use With (Application) | External thread sample tubes (T501) |
Closure Material | Polypropylene |
Quantity | 1000 Pk. |
Type | Screw Cap |
Closure Size | 1.5 to 5 mL tubes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction