Learn More
Fisherbrand™ Visitorspec Safety Glasses
Catalog No. 191302090 Shop All Fisherbrand Products
Click to view available options
:
Uncoated lens
This item is not returnable. View return policy
This item is not returnable. View return policy
Ideal for protecting visitors in industrial or medical settings.
  • Economical safety spectacles
  • 100% impact-resistant polycarbonate wraparound unilens design with blue temples
  • Clear lens absorbs >99.9% of harmful UV light
  • Uncoated lens
  • Fits over most eye-glasses
Compliance
Meet ANSI™ Z87.1-2010 standards.
Specifications
Lens Coating Uncoated
Certifications/Compliance Meets ANSI Z87.1-2003 standards.
Frame Color Blue
Temple Style Non-adjustable-length
Lens Material Polycarbonate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.